Learn More
Medchemexpress LLC DSPE-PEG-Folate, MW 3350 | 1236288-25-7 | 98.2% | C65H114N11O16P | 100 MG
SDP
Catalog No. 5000354960
Encompass_Preferred
DSPE-PEG-Folate, MW 3350 is a PEG derivative that incorporates folic acid. This compound is designed with a targeting effect, allowing it to bind to folate receptors found in cancer cells. It can form micelles or lipid bilayers, making it suitable for research in targeted drug delivery systems.
  • PEG derivative incorporating folic acid
  • Targets folate receptors in cancer cells
  • Capable of forming micelles or lipid bilayers
  • Suitable for targeted drug delivery system research
  • Nanoparticles enter cells via FA receptor-mediated endocytosis
  • Increases drug elimination half-life
  • Prolongs blood circulation time
Catalog No. 50-003-54960 Supplier Medchemexpress LLC Supplier No. HY144009100MG
DSPE-PEG-Folate, MW 3350 is a PEG derivative that incorporates folic acid. This compound is designed with a targeting effect, allowing it to bind to folate receptors found in cancer cells. It can form micelles or lipid bilayers, making it suitable for research in targeted drug delivery systems.
  • PEG derivative incorporating folic acid
  • Targets folate receptors in cancer cells
  • Capable of forming micelles or lipid bilayers
  • Suitable for targeted drug delivery system research
  • Nanoparticles enter cells via FA receptor-mediated endocytosis
  • Increases drug elimination half-life
  • Prolongs blood circulation time