Learn More
Medchemexpress LLC DSPE-PEG-Maleimide | 1612754-08-1 | 100.0% | 2000 | 50 MG
SDP
Catalog No. 5000394649
Encompass_Preferred
DSPE-PEG-Maleimide (MW 2000) is a functional lipid component and a thiol-reactive crosslinker. It undergoes Michael addition with the thiol groups of thiolated or cyclic RGD peptides to form stable thioether bonds and DSPE-PEG (2000)-RGD. This product is applicable to research on drug delivery.
  • Functional lipid component
  • Thiol-reactive crosslinker
  • Undergoes Michael addition with thiol groups
  • Forms stable thioether bonds
  • Forms DSPE-PEG (2000)-RGD with thiolated or cyclic RGD peptides
  • Applicable to research on drug delivery
Catalog No. 50-003-94649 Supplier Medchemexpress LLC Supplier No. HY16664850MG
DSPE-PEG-Maleimide (MW 2000) is a functional lipid component and a thiol-reactive crosslinker. It undergoes Michael addition with the thiol groups of thiolated or cyclic RGD peptides to form stable thioether bonds and DSPE-PEG (2000)-RGD. This product is applicable to research on drug delivery.
  • Functional lipid component
  • Thiol-reactive crosslinker
  • Undergoes Michael addition with thiol groups
  • Forms stable thioether bonds
  • Forms DSPE-PEG (2000)-RGD with thiolated or cyclic RGD peptides
  • Applicable to research on drug delivery