Learn More
Medchemexpress LLC DSPE-PEG-Maleimide | 99.8% | 50 MG
SDP
Catalog No. 5000353794
Encompass_Preferred
DSPE-PEG-Maleimide, MW 5000, is a linker lipid that contains DSPE phospholipid and Maleimide. It serves as a crucial component in the synthesis of various liposomal formulations, including HELDC liposomes, sterically stabilized liposomes, and immunoliposomes.
  • Used as a linker lipid in immunoliposome synthesis.
  • Can be coupled with thiol-p5314-29 peptide.
  • Sterically stabilized liposomes containing DSPE-PEG-maleimide can be coupled with thiolated monoclonal antibody OX26 (OX26-SH).
  • HELDC liposomes prepared with 3% DSPE-PEG-maleimide show no change in particle size over two weeks, with an average diameter of 120 nm.
Catalog No. 50-003-53794 Supplier Medchemexpress LLC Supplier No. HY14074050MG
DSPE-PEG-Maleimide, MW 5000, is a linker lipid that contains DSPE phospholipid and Maleimide. It serves as a crucial component in the synthesis of various liposomal formulations, including HELDC liposomes, sterically stabilized liposomes, and immunoliposomes.
  • Used as a linker lipid in immunoliposome synthesis.
  • Can be coupled with thiol-p5314-29 peptide.
  • Sterically stabilized liposomes containing DSPE-PEG-maleimide can be coupled with thiolated monoclonal antibody OX26 (OX26-SH).
  • HELDC liposomes prepared with 3% DSPE-PEG-maleimide show no change in particle size over two weeks, with an average diameter of 120 nm.