Learn More
Medchemexpress LLC DSPE-PEG-maleimide (MW 3400) ammonium | ≥95.0% | 3400 g/mol | (C2H4O)nC52H93N3NaO13P | 500 MG
SDP
Catalog No. 5000281763
Encompass_Preferred
DSPE-PEG-Maleimide (MW 3400) ammonium is a DSPE-based phospholipid conjugated to a polyethylene glycol chain (approx. 3.4 kDa) terminating in a maleimide reactive group. It is used to incorporate reactive PEGylated lipids into liposomes, nanostructured lipid carriers, and other nanoparticle systems to enable thiol-specific conjugation and improve colloidal stability.
Catalog No. 50-002-81763 Supplier Medchemexpress LLC Supplier No. HYW441009A500MG
DSPE-PEG-Maleimide (MW 3400) ammonium is a DSPE-based phospholipid conjugated to a polyethylene glycol chain (approx. 3.4 kDa) terminating in a maleimide reactive group. It is used to incorporate reactive PEGylated lipids into liposomes, nanostructured lipid carriers, and other nanoparticle systems to enable thiol-specific conjugation and improve colloidal stability.