Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dspe-peg-maleimide, MW 5000 | 99.8% | 100 MG
SDP

Catalog No. 5000355497
Encompass_Preferred

Dspe-peg-maleimide, MW 5000, is a linker lipid containing DSPE phospholipid and Maleimide. This compound is used in the synthesis of various liposomes and as a linker in immunoliposome synthesis for laboratory applications.

  • Contains DSPE phospholipid and Maleimide.
  • Utilized in liposome synthesis (HELDC, sterically stabilized, and immunoliposomes).
  • Can be coupled with thiol-p5314-29 peptide for immunoliposome synthesis.
  • Sterically stabilized liposomes can be coupled with thiolated monoclonal antibody OX26 (OX26-SH).
  • HELDC liposomes with 3% DSPE-PEG-maleimide maintain particle size stability over two weeks (avg. 120 nm diameter).
  • Molecular weight: 5000 (average).
  • Appears as a white to off-white solid.
  • For research use only.

Catalog No. 50-003-55497 Supplier Medchemexpress LLC Supplier No. HY140740100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dspe-peg-maleimide, MW 5000, is a linker lipid containing DSPE phospholipid and Maleimide. This compound is used in the synthesis of various liposomes and as a linker in immunoliposome synthesis for laboratory applications.

  • Contains DSPE phospholipid and Maleimide.
  • Utilized in liposome synthesis (HELDC, sterically stabilized, and immunoliposomes).
  • Can be coupled with thiol-p5314-29 peptide for immunoliposome synthesis.
  • Sterically stabilized liposomes can be coupled with thiolated monoclonal antibody OX26 (OX26-SH).
  • HELDC liposomes with 3% DSPE-PEG-maleimide maintain particle size stability over two weeks (avg. 120 nm diameter).
  • Molecular weight: 5000 (average).
  • Appears as a white to off-white solid.
  • For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.