Learn More
Broadpharm DSPE-PEG-Mannose, MW 5000, Unit Size 100 mg
Catalog No. NC2759188
Encompass
DSPE-PEG-Mannose is a high-purity polyethylene glycol (PEG) polymer engineered for advanced scientific applications. This amphiphilic conjugate consists of a hydrophobic DSPE moiety and a hydrophilic mannose-terminated PEG chain, which facilitates the encapsulation of hydrophobic drugs while enhancing water solubility for efficient drug delivery. Its broad utility spans medical research, drug release studies, nanotechnology, and the development of targeted drug carriers such as liposomes. This versatile compound significantly improves the selectivity of PEGylation reactions and supports various applications including peptide synthesis and the creation of functional coatings.
  • Molecular weight of 5000
  • Unit size of 100 mg
  • Hydrophobic DSPE and hydrophilic PEG components
  • Increases selectivity of PEGylation reactions
  • Improved solubility due to D-glucose
  • Utilized in drug delivery systems and mRNA vaccines
Catalog No. NC2759188 Supplier Broadpharm Supplier No. BP28050100MG
DSPE-PEG-Mannose is a high-purity polyethylene glycol (PEG) polymer engineered for advanced scientific applications. This amphiphilic conjugate consists of a hydrophobic DSPE moiety and a hydrophilic mannose-terminated PEG chain, which facilitates the encapsulation of hydrophobic drugs while enhancing water solubility for efficient drug delivery. Its broad utility spans medical research, drug release studies, nanotechnology, and the development of targeted drug carriers such as liposomes. This versatile compound significantly improves the selectivity of PEGylation reactions and supports various applications including peptide synthesis and the creation of functional coatings.
  • Molecular weight of 5000
  • Unit size of 100 mg
  • Hydrophobic DSPE and hydrophilic PEG components
  • Increases selectivity of PEGylation reactions
  • Improved solubility due to D-glucose
  • Utilized in drug delivery systems and mRNA vaccines