Learn More
Medchemexpress LLC Dspe-peg-nhs | MFCD03098978 | 80.0% | 5000 (average) | 100 MG
SDP
Catalog No. 5000281091
Encompass_Preferred
DSPE-PEG-NHS (average molecular weight 5000) is a PEG-modified phospholipid bearing an N-hydroxysuccinimide (NHS) ester. It functions as an amphiphilic linker reagent for covalent attachment of primary amine-containing ligands to lipid surfaces, enabling targeted modification of liposomes and other nanoparticle systems used in drug delivery research.
  • Amphiphilic PEG-lipid suitable for liposome and nanoparticle surface functionalization.
  • NHS ester enables efficient covalent coupling to primary amines under mild conditions.
  • Average molecular weight 5000 provides extended hydrophilicity and steric stabilization.
  • Supplied as a powder, stable under recommended frozen storage for long-term use.
  • Useful for preparing targeted delivery systems and conjugating biomolecules.
  • Purity approximately 80.0% as provided by the manufacturer.
Catalog No. 50-002-81091 Supplier Medchemexpress LLC Supplier No. HYW441016100MG
DSPE-PEG-NHS (average molecular weight 5000) is a PEG-modified phospholipid bearing an N-hydroxysuccinimide (NHS) ester. It functions as an amphiphilic linker reagent for covalent attachment of primary amine-containing ligands to lipid surfaces, enabling targeted modification of liposomes and other nanoparticle systems used in drug delivery research.
  • Amphiphilic PEG-lipid suitable for liposome and nanoparticle surface functionalization.
  • NHS ester enables efficient covalent coupling to primary amines under mild conditions.
  • Average molecular weight 5000 provides extended hydrophilicity and steric stabilization.
  • Supplied as a powder, stable under recommended frozen storage for long-term use.
  • Useful for preparing targeted delivery systems and conjugating biomolecules.
  • Purity approximately 80.0% as provided by the manufacturer.