Learn More
Medchemexpress LLC Dspe-PEG-NHS (N-hydroxysuccinimide ester of DSPE-PEG) | 98.0% | 2000 (average) | 100 MG
SDP
Catalog No. 5000280970
Encompass_Preferred
DSPE-PEG-NHS (average molecular weight 2000) is an NHS-activated PEGylated phospholipid used to introduce polyethylene glycol chains onto lipid-based nanoparticles and to conjugate amine-containing ligands via formation of stable amide bonds. It is commonly applied in liposome and micelle formulation, surface PEGylation, and ligand attachment for targeted delivery. Typical purity is 98%.
  • NHS ester for efficient amine coupling reactions
  • Average PEG molecular weight 2000 for steric stabilization
  • Compatible with liposome and micelle formulation workflows
  • Enables surface PEGylation and conjugation of targeting ligands
  • High purity suitable for research applications
  • Available as a dry powder for convenient storage and handling
Catalog No. 50-002-80970 Supplier Medchemexpress LLC Supplier No. HYW441014100MG
DSPE-PEG-NHS (average molecular weight 2000) is an NHS-activated PEGylated phospholipid used to introduce polyethylene glycol chains onto lipid-based nanoparticles and to conjugate amine-containing ligands via formation of stable amide bonds. It is commonly applied in liposome and micelle formulation, surface PEGylation, and ligand attachment for targeted delivery. Typical purity is 98%.
  • NHS ester for efficient amine coupling reactions
  • Average PEG molecular weight 2000 for steric stabilization
  • Compatible with liposome and micelle formulation workflows
  • Enables surface PEGylation and conjugation of targeting ligands
  • High purity suitable for research applications
  • Available as a dry powder for convenient storage and handling