Learn More
Medchemexpress LLC Dspe-peg-oh | 99.89% | 100 MG
SDP
Catalog No. 5000353541
Encompass_Preferred
DSPE-PEG-OH (MW 2000) is a hydroxyl-terminated PEGylated phospholipid. Its hydrophobic tail allows for the encapsulation and aggregation of other hydrophobic drugs, and its hydroxyl terminus is suitable for further reactions. DSPE-PEG-OH (MW 2000) can be used to prepare liposomes or lipid nanoparticles.
  • Hydroxyl-terminated PEGylated phospholipid.
  • Hydrophobic tail enables encapsulation and aggregation of other hydrophobic drugs.
  • Hydroxyl terminus suitable for further reactions.
  • Can be used to prepare liposomes or lipid nanoparticles.
Catalog No. 50-003-53541 Supplier Medchemexpress LLC Supplier No. HY140741100MG
DSPE-PEG-OH (MW 2000) is a hydroxyl-terminated PEGylated phospholipid. Its hydrophobic tail allows for the encapsulation and aggregation of other hydrophobic drugs, and its hydroxyl terminus is suitable for further reactions. DSPE-PEG-OH (MW 2000) can be used to prepare liposomes or lipid nanoparticles.
  • Hydroxyl-terminated PEGylated phospholipid.
  • Hydrophobic tail enables encapsulation and aggregation of other hydrophobic drugs.
  • Hydroxyl terminus suitable for further reactions.
  • Can be used to prepare liposomes or lipid nanoparticles.