Learn More
Medchemexpress LLC Dspe-peg-oh (mw 2000) | 892144-24-0 | 99.9% | (C2H4O)nC42H82NO10P | 25 MG
SDP
Catalog No. 5000352663
Encompass_Preferred
DSPE-PEG-OH (MW 2000) is a hydroxyl-terminated PEGylated phospholipid. Its hydrophobic tail allows for the encapsulation and aggregation of other hydrophobic drugs, and the hydroxyl terminus is suitable for further reactions. This product can be used to prepare liposomes or lipid nanoparticles.
  • Hydroxyl-terminated PEGylated phospholipid
  • Hydrophobic tail allows encapsulation and aggregation of other hydrophobic drugs
  • Hydroxyl terminus is suitable for further reactions
  • Can be used to prepare liposomes or lipid nanoparticles
Catalog No. 50-003-52663 Supplier Medchemexpress LLC Supplier No. 25MGHY140741
DSPE-PEG-OH (MW 2000) is a hydroxyl-terminated PEGylated phospholipid. Its hydrophobic tail allows for the encapsulation and aggregation of other hydrophobic drugs, and the hydroxyl terminus is suitable for further reactions. This product can be used to prepare liposomes or lipid nanoparticles.
  • Hydroxyl-terminated PEGylated phospholipid
  • Hydrophobic tail allows encapsulation and aggregation of other hydrophobic drugs
  • Hydroxyl terminus is suitable for further reactions
  • Can be used to prepare liposomes or lipid nanoparticles