Learn More
Medchemexpress LLC DSPE-PEG-SH | 1462982-18-8 | 90.0% | 1,000 Da | C44H86NO10PS | 250 MG
SDP
Catalog No. 5000281173
Encompass_Preferred
DSPE-PEG-SH (MW 1000) is a thiol-terminated PEGylated phospholipid reagent used to introduce free thiol groups onto liposome or nanoparticle surfaces for subsequent bioconjugation. It is supplied for research use in small laboratory pack sizes and is commonly employed in formulation, surface modification, and targeted delivery workflows.
  • Thiol functional group for conjugation to maleimide reagents.
  • Average molecular weight of approximately 1,000 Da.
  • Amphiphilic phospholipid-PEG structure for incorporation into lipid bilayers.
  • Enables surface modification of liposomes and nanoparticles.
  • Supplied in small research pack sizes for laboratory use.
  • Typical reported purity around 90%.
Catalog No. 50-002-81173 Supplier Medchemexpress LLC Supplier No. HYW440895250MG
DSPE-PEG-SH (MW 1000) is a thiol-terminated PEGylated phospholipid reagent used to introduce free thiol groups onto liposome or nanoparticle surfaces for subsequent bioconjugation. It is supplied for research use in small laboratory pack sizes and is commonly employed in formulation, surface modification, and targeted delivery workflows.
  • Thiol functional group for conjugation to maleimide reagents.
  • Average molecular weight of approximately 1,000 Da.
  • Amphiphilic phospholipid-PEG structure for incorporation into lipid bilayers.
  • Enables surface modification of liposomes and nanoparticles.
  • Supplied in small research pack sizes for laboratory use.
  • Typical reported purity around 90%.