Learn More
Medchemexpress LLC Dspe-PEG-SH | 90.0% | 1,000 Da | 50 MG
SDP
Catalog No. 5000281325
Encompass_Preferred
Thiol-terminated DSPE-PEG-SH (MW 1000) is an amphiphilic pegylated phospholipid composed of a DSPE lipid anchor linked to an approximately 1,000 Da polyethylene glycol chain that ends in a sulfhydryl group. It is intended for formulation of PEGylated liposomes and lipid nanoparticles and for surface functionalization or bioconjugation using thiol-specific chemistries.
  • Thiol terminal for conjugation to gold, maleimide, or crosslinkers.
  • Amphiphilic structure suitable for liposome and nanoparticle formulation.
  • PEG chain ~1,000 Da provides steric stabilization and improved circulation.
  • Typical purity around 90.0% as supplied.
  • Soluble in common organic solvents used for lipid formulation.
  • Packaged in small research quantities for laboratory use.
Catalog No. 50-002-81325 Supplier Medchemexpress LLC Supplier No. HYW44089550MG
Thiol-terminated DSPE-PEG-SH (MW 1000) is an amphiphilic pegylated phospholipid composed of a DSPE lipid anchor linked to an approximately 1,000 Da polyethylene glycol chain that ends in a sulfhydryl group. It is intended for formulation of PEGylated liposomes and lipid nanoparticles and for surface functionalization or bioconjugation using thiol-specific chemistries.
  • Thiol terminal for conjugation to gold, maleimide, or crosslinkers.
  • Amphiphilic structure suitable for liposome and nanoparticle formulation.
  • PEG chain ~1,000 Da provides steric stabilization and improved circulation.
  • Typical purity around 90.0% as supplied.
  • Soluble in common organic solvents used for lipid formulation.
  • Packaged in small research quantities for laboratory use.