Learn More
Medchemexpress LLC DSPE-PEG-SH | 90.0% | 1,000 Da | C44H86NO10PS | 100 MG
SDP
Catalog No. 5000280500
Encompass_Preferred
DSPE-PEG-SH (MW 1000) is a thiol-terminated pegylated phospholipid composed of a DSPE lipid anchor linked to an approximately 1,000 Da polyethylene glycol chain ending in a free sulfhydryl. The amphiphilic molecule integrates into lipid bilayers and is used to functionalize liposomes and nanoparticles and to enable covalent conjugation to maleimide-activated ligands.
  • Provides a thiol functional group for covalent conjugation.
  • Anchors into lipid bilayers via the DSPE lipid tail.
  • Enhances colloidal stability and circulation through PEGylation.
  • Suitable for surface functionalization of liposomes and nanoparticles.
  • Average molecular weight approximately 1,000 Da.
  • Typical purity around 90%.
Catalog No. 50-002-80500 Supplier Medchemexpress LLC Supplier No. HYW440895100MG
DSPE-PEG-SH (MW 1000) is a thiol-terminated pegylated phospholipid composed of a DSPE lipid anchor linked to an approximately 1,000 Da polyethylene glycol chain ending in a free sulfhydryl. The amphiphilic molecule integrates into lipid bilayers and is used to functionalize liposomes and nanoparticles and to enable covalent conjugation to maleimide-activated ligands.
  • Provides a thiol functional group for covalent conjugation.
  • Anchors into lipid bilayers via the DSPE lipid tail.
  • Enhances colloidal stability and circulation through PEGylation.
  • Suitable for surface functionalization of liposomes and nanoparticles.
  • Average molecular weight approximately 1,000 Da.
  • Typical purity around 90%.