Learn More
Medchemexpress LLC Dspe-peg-sh | 99.4% | 2000 (average) | (C2H4O)nC45H87N2O10PS | 25 MG
SDP
Catalog No. 5000281461
Encompass_Preferred
DSPE-PEG-SH is a thiolated pegylated phospholipid (average MW 2000) that provides a terminal -SH for covalent surface modification of lipid assemblies. Its amphipathic DSPE-PEG structure enables incorporation into liposomes and lipid nanoparticles for ligand attachment, stabilization, and targeted delivery via formation of stable thioether bonds with maleimide-functionalized molecules.
  • Provides a terminal thiol for covalent conjugation.
  • Incorporates into lipid bilayers for surface functionalization.
  • Enables stable thioether linkage with maleimide reagents.
  • Suitable for liposome and lipid nanoparticle formulations.
  • Facilitates targeted delivery via ligand attachment.
  • Reported high purity supports reproducible formulation.
  • Soluble in water and DMSO with warming and sonication.
Catalog No. 50-002-81461 Supplier Medchemexpress LLC Supplier No. HYW44089625MG
DSPE-PEG-SH is a thiolated pegylated phospholipid (average MW 2000) that provides a terminal -SH for covalent surface modification of lipid assemblies. Its amphipathic DSPE-PEG structure enables incorporation into liposomes and lipid nanoparticles for ligand attachment, stabilization, and targeted delivery via formation of stable thioether bonds with maleimide-functionalized molecules.
  • Provides a terminal thiol for covalent conjugation.
  • Incorporates into lipid bilayers for surface functionalization.
  • Enables stable thioether linkage with maleimide reagents.
  • Suitable for liposome and lipid nanoparticle formulations.
  • Facilitates targeted delivery via ligand attachment.
  • Reported high purity supports reproducible formulation.
  • Soluble in water and DMSO with warming and sonication.