Learn More
Broadpharm DSPE-PEG-TCO, 2000 Da, 25 mg, Solid, ≥90% Purity, -20°C Storage, Ambient Shipping, In Stock
Catalog No. NC2818074
Encompass
DSPE-PEG-TCO, MW 2000, is a functionalized PEG-lipid conjugate that facilitates micelle formation in aqueous media and is integral to lipid nanoparticle formulations for drug delivery. This product enables highly efficient bioorthogonal labeling via click chemistry with tetrazine-containing compounds, designed to improve the delivery efficiency and tissue specificity of poorly soluble drugs in research applications.
  • Enhanced drug delivery for poorly soluble drugs.
  • Bioorthogonal labeling with tetrazine-containing compounds.
  • Facilitates micelle formation in aqueous solutions.
  • Utilized in lipid nanoparticle formulations.
  • Stable under recommended storage conditions.
Catalog No. NC2818074 Supplier Broadpharm Supplier No. BP4010225MG
DSPE-PEG-TCO, MW 2000, is a functionalized PEG-lipid conjugate that facilitates micelle formation in aqueous media and is integral to lipid nanoparticle formulations for drug delivery. This product enables highly efficient bioorthogonal labeling via click chemistry with tetrazine-containing compounds, designed to improve the delivery efficiency and tissue specificity of poorly soluble drugs in research applications.
  • Enhanced drug delivery for poorly soluble drugs.
  • Bioorthogonal labeling with tetrazine-containing compounds.
  • Facilitates micelle formation in aqueous solutions.
  • Utilized in lipid nanoparticle formulations.
  • Stable under recommended storage conditions.