Learn More
Medchemexpress LLC DSPE-PEG2-mal | 1915739-87-5 | 99.9% | 1058.37 | C55H100N3O14P | 5 MG
SDP
Catalog No. 5000221875
Encompass_Preferred
DSPE-PEG2-mal is a PEG-based maleimide-functionalized linker that couples a DSPE lipid moiety to a short PEG spacer terminated with a maleimide group. It is intended for thiol-directed conjugation in the synthesis of PROTACs and other bioconjugates, providing a lipid anchor with improved solubility and membrane association. Manufacturer-reported properties include high purity, molecular weight ~1058.37, and formula C55H100N3O14P.
  • Provides a maleimide terminus for thiol-specific conjugation.
  • Contains a DSPE lipid moiety for membrane anchoring.
  • Includes a short PEG spacer to improve solubility and reduce steric hindrance.
  • High reported purity suitable for research applications.
  • Solid, white to off-white appearance for convenient handling.
Catalog No. 50-002-21875 Supplier Medchemexpress LLC Supplier No. HY1347455MG
DSPE-PEG2-mal is a PEG-based maleimide-functionalized linker that couples a DSPE lipid moiety to a short PEG spacer terminated with a maleimide group. It is intended for thiol-directed conjugation in the synthesis of PROTACs and other bioconjugates, providing a lipid anchor with improved solubility and membrane association. Manufacturer-reported properties include high purity, molecular weight ~1058.37, and formula C55H100N3O14P.
  • Provides a maleimide terminus for thiol-specific conjugation.
  • Contains a DSPE lipid moiety for membrane anchoring.
  • Includes a short PEG spacer to improve solubility and reduce steric hindrance.
  • High reported purity suitable for research applications.
  • Solid, white to off-white appearance for convenient handling.