Learn More
Medchemexpress LLC Dspe-peg2-mal | 1915739-87-5 | 99.9% | C55H100N3O14P | 10MG
SDP
Catalog No. 5000259230
Encompass_Preferred
DSPE-PEG2-mal is a PEG-based linker comprising a distearoylphosphatidylethanolamine (DSPE) lipid connected via a short PEG2 spacer to a maleimide functional group. It is intended for thiol-selective conjugation and linker installation in PROTACs, lipidated probes, and general bioconjugation research.
  • Contains a maleimide terminus for selective reaction with thiols.
  • Features a short PEG2 spacer to enhance solubility and flexibility.
  • Includes a DSPE lipid moiety for membrane anchoring and lipidation.
  • High reported purity supports reproducible synthesis and analysis.
  • Solid, white to off-white form for ease of handling and storage.
  • Available in multiple small-scale pack sizes suitable for research use.
Catalog No. 50-002-59230 Supplier Medchemexpress LLC Supplier No. HY13474510MG
DSPE-PEG2-mal is a PEG-based linker comprising a distearoylphosphatidylethanolamine (DSPE) lipid connected via a short PEG2 spacer to a maleimide functional group. It is intended for thiol-selective conjugation and linker installation in PROTACs, lipidated probes, and general bioconjugation research.
  • Contains a maleimide terminus for selective reaction with thiols.
  • Features a short PEG2 spacer to enhance solubility and flexibility.
  • Includes a DSPE lipid moiety for membrane anchoring and lipidation.
  • High reported purity supports reproducible synthesis and analysis.
  • Solid, white to off-white form for ease of handling and storage.
  • Available in multiple small-scale pack sizes suitable for research use.