Learn More
Medchemexpress LLC DSPE-PEG2000-COOH | 1445832-74-5 | 99.9% | 10 MG
SDP
Catalog No. 5000295087
Encompass_Preferred
DSPE-PEG(2000)-COOH is a PEGylated derivative of 1,2-distearoyl-sn-glycerol-3-phosphorylethanolamine featuring a carboxylic acid-terminated polyethylene glycol chain of average molecular weight ~2000. It is provided as a solid for use in liposome and lipid nanoparticle formulations where surface PEGylation and terminal functionalization are required. Store powder at -20°C and handle according to supplied safety documentation.
  • Provides surface PEGylation to increase circulation time in vivo.
  • Terminal carboxyl group enables covalent conjugation and further functionalization.
  • Suitable for preparation of liposomes and lipid nanoparticles.
  • Average molecular weight gives consistent PEG chain length and steric stabilization.
  • Solid form with defined storage conditions for stability.
Catalog No. 50-002-95087 Supplier Medchemexpress LLC Supplier No. HY17070610MG
DSPE-PEG(2000)-COOH is a PEGylated derivative of 1,2-distearoyl-sn-glycerol-3-phosphorylethanolamine featuring a carboxylic acid-terminated polyethylene glycol chain of average molecular weight ~2000. It is provided as a solid for use in liposome and lipid nanoparticle formulations where surface PEGylation and terminal functionalization are required. Store powder at -20°C and handle according to supplied safety documentation.
  • Provides surface PEGylation to increase circulation time in vivo.
  • Terminal carboxyl group enables covalent conjugation and further functionalization.
  • Suitable for preparation of liposomes and lipid nanoparticles.
  • Average molecular weight gives consistent PEG chain length and steric stabilization.
  • Solid form with defined storage conditions for stability.