Learn More
Medchemexpress LLC DSPE-PEG2000-cRGD | 96.2% | 10 MG
SDP
Catalog No. 5000295382
Encompass_Preferred
DSPE-PEG2000-cRGD is a polyethylene glycol-lipid conjugate that links DSPE to a PEG2000 spacer terminated with a cyclic RGD peptide targeting αvβ3 integrins. Supplied as a white to off-white solid for research use, it is intended for incorporation into lipid nanoparticles and other targeted delivery systems to enhance tumor and neovasculature binding.
  • Provides αvβ3-targeting through a cRGD ligand.
  • Facilitates surface functionalization of lipid nanoparticles.
  • PEG2000 spacer improves circulation and steric stability.
  • Supplied as a solid with reported purity of ~96.2%.
  • Stable under frozen storage; follow supplier storage instructions.
Catalog No. 50-002-95382 Supplier Medchemexpress LLC Supplier No. HY17246410MG
DSPE-PEG2000-cRGD is a polyethylene glycol-lipid conjugate that links DSPE to a PEG2000 spacer terminated with a cyclic RGD peptide targeting αvβ3 integrins. Supplied as a white to off-white solid for research use, it is intended for incorporation into lipid nanoparticles and other targeted delivery systems to enhance tumor and neovasculature binding.
  • Provides αvβ3-targeting through a cRGD ligand.
  • Facilitates surface functionalization of lipid nanoparticles.
  • PEG2000 spacer improves circulation and steric stability.
  • Supplied as a solid with reported purity of ~96.2%.
  • Stable under frozen storage; follow supplier storage instructions.