Learn More
Medchemexpress LLC DSPE-PEG2000-cRGD | 96.2% | 2000 | 25 MG
SDP
Catalog No. 5000384788
Encompass_Preferred
DSPE-PEG2000-cRGD is a PEG compound composed of DSPE and an αvβ3 targeting peptide (cRGD). The cRGD peptide specifically binds to αvβ3 on the surface of many cancer cells and neovascular cells. It can be used for drug delivery.
  • Composed of DSPE and an αvβ3 targeting peptide (cRGD)
  • cRGD peptide specifically binds to αvβ3 on the surface of many cancer cells and neovascular cells
  • Can be used for drug delivery
  • Can be used to construct pp-siRNA/CLD/DNCA/DSPE-PEG2000-cRGD (PCNR) nanoparticles that specifically target tumor cells that highly express integrin αvβ3
  • Incorporating it into liposomes can form spatially stable and integrin-targeting liposomes
  • Can be used to construct C-siRNA/MTSLR-U and C-siRNA/MTSLR, which have anti-hepatocellular carcinoma activity
Catalog No. 50-003-84788 Supplier Medchemexpress LLC Supplier No. 25MGHY172464
DSPE-PEG2000-cRGD is a PEG compound composed of DSPE and an αvβ3 targeting peptide (cRGD). The cRGD peptide specifically binds to αvβ3 on the surface of many cancer cells and neovascular cells. It can be used for drug delivery.
  • Composed of DSPE and an αvβ3 targeting peptide (cRGD)
  • cRGD peptide specifically binds to αvβ3 on the surface of many cancer cells and neovascular cells
  • Can be used for drug delivery
  • Can be used to construct pp-siRNA/CLD/DNCA/DSPE-PEG2000-cRGD (PCNR) nanoparticles that specifically target tumor cells that highly express integrin αvβ3
  • Incorporating it into liposomes can form spatially stable and integrin-targeting liposomes
  • Can be used to construct C-siRNA/MTSLR-U and C-siRNA/MTSLR, which have anti-hepatocellular carcinoma activity