Learn More
Medchemexpress LLC Dspe-peg2000-crgdkgpdc (disulfide bridge:Cys1-Cys9) | 95.0% | 2000 (Average) | 25 MG
SDP
Catalog No. 5000385160
Encompass_Preferred
DSPE-PEG2000-iRGD is a PEG compound composed of DSPE and an αv-integrins targeting peptide (iRGD). The iRGD peptide binds to αv-integrins and is then proteolytically cleaved in the tumor to produce CRGDK/R, which interacts with neuropilin-1. This compound has tumor-targeting and tumor-penetrating properties and can be used for agent delivery.
  • Irgd peptide binds to αv-integrins.
  • Proteolytically cleaved in the tumor to produce CRGDK/R to interact with neuropilin-1.
  • Possesses tumor-targeting and tumor-penetrating properties.
  • Can be used for agent delivery.
  • Used to construct nanomicelles for targeted delivery of salinomycin to intervene in liver cancer cells and cancer stem cells.
  • Used to construct a tumor-penetrating peptide liposome that exhibits antitumor effects, including a significant reduction in tumor volume and an increase in body weight and spleen index.
Catalog No. 50-003-85160 Supplier Medchemexpress LLC Supplier No. 25MGHY172494
DSPE-PEG2000-iRGD is a PEG compound composed of DSPE and an αv-integrins targeting peptide (iRGD). The iRGD peptide binds to αv-integrins and is then proteolytically cleaved in the tumor to produce CRGDK/R, which interacts with neuropilin-1. This compound has tumor-targeting and tumor-penetrating properties and can be used for agent delivery.
  • Irgd peptide binds to αv-integrins.
  • Proteolytically cleaved in the tumor to produce CRGDK/R to interact with neuropilin-1.
  • Possesses tumor-targeting and tumor-penetrating properties.
  • Can be used for agent delivery.
  • Used to construct nanomicelles for targeted delivery of salinomycin to intervene in liver cancer cells and cancer stem cells.
  • Used to construct a tumor-penetrating peptide liposome that exhibits antitumor effects, including a significant reduction in tumor volume and an increase in body weight and spleen index.