Learn More
Medchemexpress LLC DSPE-PEG2000-Cys-SRNLIDC (disulfide bridge: Cys1-Cys8) | 2000 (Average) | 5 MG
SDP
Catalog No. 5000385325
Encompass_Preferred
DSPE-PEG2000-Cys-SRNLIDC is a PEG compound composed of DSPE and a cyclic oligopeptide (pPB). The pPB component exhibits a strong binding affinity with PDGFRβ, which is overexpressed on activated hepatic stellate cells (HSC). This compound is suitable for drug delivery applications.
  • Composed of DSPE and a cyclic oligopeptide (pPB)
  • pPB binds strongly with PDGFRβ
  • Suitable for drug delivery
Catalog No. 50-003-85325 Supplier Medchemexpress LLC Supplier No. HY172275A5MG
DSPE-PEG2000-Cys-SRNLIDC is a PEG compound composed of DSPE and a cyclic oligopeptide (pPB). The pPB component exhibits a strong binding affinity with PDGFRβ, which is overexpressed on activated hepatic stellate cells (HSC). This compound is suitable for drug delivery applications.
  • Composed of DSPE and a cyclic oligopeptide (pPB)
  • pPB binds strongly with PDGFRβ
  • Suitable for drug delivery