Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dspe-peg2000-cys-srnlidc (disulfide bridge cys1-cys8) | 95.2% | 2000 Da (average) | 10 MG
SDP

Catalog No. 5000295442
Encompass_Preferred

DSPE-PEG2000-Cys-SRNLIDC is a lipid-PEG conjugate that links a DSPE lipid to a cyclic SRNLIDC peptide through a PEG2000 spacer containing a disulfide bridge (Cys1-Cys8). It is provided for research use to functionalize lipid nanoparticles or liposomes for targeting and to enable disulfide-triggered intracellular release. PEGylation improves colloidal stability and compatibility with aqueous formulations.

  • Enables surface functionalization of lipid nanoparticles.
  • Provides disulfide-mediated intracellular release of linked cargo.
  • PEG2000 spacer enhances colloidal stability and circulation.
  • Supplied with high reported purity for research applications.
  • Soluble in DMSO for stock solution preparation.
  • Recommended storage at -20°C to preserve integrity.

Catalog No. 50-002-95442 Supplier Medchemexpress LLC Supplier No. HY172275A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DSPE-PEG2000-Cys-SRNLIDC is a lipid-PEG conjugate that links a DSPE lipid to a cyclic SRNLIDC peptide through a PEG2000 spacer containing a disulfide bridge (Cys1-Cys8). It is provided for research use to functionalize lipid nanoparticles or liposomes for targeting and to enable disulfide-triggered intracellular release. PEGylation improves colloidal stability and compatibility with aqueous formulations.

  • Enables surface functionalization of lipid nanoparticles.
  • Provides disulfide-mediated intracellular release of linked cargo.
  • PEG2000 spacer enhances colloidal stability and circulation.
  • Supplied with high reported purity for research applications.
  • Soluble in DMSO for stock solution preparation.
  • Recommended storage at -20°C to preserve integrity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.