Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DSPE-PEG2000-GE11 | 95.3% | 10 MG
SDP

Catalog No. 5000295383
Encompass_Preferred

DSPE-PEG2000-GE11 is a PEGylated lipid-peptide conjugate combining a DSPE lipid anchor, a PEG2000 spacer, and the EGFR-targeting GE11 peptide for targeted drug delivery and lipid nanoparticle formulation. It is designed to functionalize particle surfaces and direct therapeutic payloads to EGFR-overexpressing cells.

  • Targets EGFR-overexpressing cancer cells.
  • Provides a DSPE lipid anchor for membrane insertion.
  • Includes a PEG2000 spacer to improve circulation and reduce opsonization.
  • Contains the GE11 peptide for receptor-targeted delivery.
  • Soluble in DMSO with sonication and warming to 60°C.
  • Powder storage at -20°C; in solution store at -80°C for long-term.

Catalog No. 50-002-95383 Supplier Medchemexpress LLC Supplier No. HY17247010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DSPE-PEG2000-GE11 is a PEGylated lipid-peptide conjugate combining a DSPE lipid anchor, a PEG2000 spacer, and the EGFR-targeting GE11 peptide for targeted drug delivery and lipid nanoparticle formulation. It is designed to functionalize particle surfaces and direct therapeutic payloads to EGFR-overexpressing cells.

  • Targets EGFR-overexpressing cancer cells.
  • Provides a DSPE lipid anchor for membrane insertion.
  • Includes a PEG2000 spacer to improve circulation and reduce opsonization.
  • Contains the GE11 peptide for receptor-targeted delivery.
  • Soluble in DMSO with sonication and warming to 60°C.
  • Powder storage at -20°C; in solution store at -80°C for long-term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.