Learn More
Medchemexpress LLC DSPE-PEG2000-iRGD | 95.0% | 2000 (average) | 10 MG
SDP
Catalog No. 5000295384
Encompass_Preferred
DSPE-PEG2000-iRGD is a lipid-PEG conjugate formed by linking distearoylphosphatidylethanolamine to the tumor-penetrating iRGD peptide (CRGDKGPDC) via a PEG2000 spacer. Supplied as a powder, it is intended for surface functionalization of liposomes and nanoparticles to confer tumor-homing and tumor-penetrating properties for targeted drug delivery studies.
  • Provides tumor-targeting via αv integrin binding.
  • Promotes tumor penetration through neuropilin-1 interaction after proteolytic cleavage.
  • Enables surface modification of liposomes and nanoparticles for targeted delivery.
  • Contains a PEG2000 spacer for steric stabilization and extended circulation.
  • Characterized by high purity and an average molecular weight for reproducible formulation.
Catalog No. 50-002-95384 Supplier Medchemexpress LLC Supplier No. HY17249410MG
DSPE-PEG2000-iRGD is a lipid-PEG conjugate formed by linking distearoylphosphatidylethanolamine to the tumor-penetrating iRGD peptide (CRGDKGPDC) via a PEG2000 spacer. Supplied as a powder, it is intended for surface functionalization of liposomes and nanoparticles to confer tumor-homing and tumor-penetrating properties for targeted drug delivery studies.
  • Provides tumor-targeting via αv integrin binding.
  • Promotes tumor penetration through neuropilin-1 interaction after proteolytic cleavage.
  • Enables surface modification of liposomes and nanoparticles for targeted delivery.
  • Contains a PEG2000 spacer for steric stabilization and extended circulation.
  • Characterized by high purity and an average molecular weight for reproducible formulation.