Learn More
Medchemexpress LLC DSPE-PEG2000-Mal ammonium | 474922-22-0 | >99.9% | (C2H4O)nC51H92N1O13P.H3N | 25 MG
SDP
Catalog No. 5000356528
Encompass_Preferred
DSPE-PEG2000-Mal ammonium contains DSPE phospholipid and maleimide, designed for preparing nanostructured lipid carriers. It is known to extend blood circulation time and increase stability for encapsulated agents. Immune liposomes constructed with this product can be utilized for brain drug delivery.
  • Contains DSPE phospholipid and maleimide
  • Prepares nanostructured lipid carriers
  • Extends blood circulation time
  • Increases stability for encapsulated agents
  • Can be used for brain drug delivery with immune liposomes
Catalog No. 50-003-56528 Supplier Medchemexpress LLC Supplier No. 25MGHY144004
DSPE-PEG2000-Mal ammonium contains DSPE phospholipid and maleimide, designed for preparing nanostructured lipid carriers. It is known to extend blood circulation time and increase stability for encapsulated agents. Immune liposomes constructed with this product can be utilized for brain drug delivery.
  • Contains DSPE phospholipid and maleimide
  • Prepares nanostructured lipid carriers
  • Extends blood circulation time
  • Increases stability for encapsulated agents
  • Can be used for brain drug delivery with immune liposomes