Learn More
Medchemexpress LLC Dspe-peg2000-mal ammonium | 474922-22-0 | MFCD00040685 | 99.9% | (C2H4O)nC51H92N1O13P.H3N | 100 MG
SDP
Catalog No. 5000354289
Encompass_Preferred
DSPE-PEG-Maleimide contains DSPE phospholipid and maleimide for preparing nanostructured lipid carriers. It extends blood circulation time and provides higher stability for encapsulated agents. It is for research use only and not sold to patients. Immune liposomes constructed by DSPE-PEG-Maleimide can be used for the delivery of brain drugs.
  • DSPE phospholipid and maleimide for nanostructured lipid carriers
  • Extends blood circulation time
  • Provides higher stability for encapsulated agents
  • Used for delivery of brain drugs
Catalog No. 50-003-54289 Supplier Medchemexpress LLC Supplier No. HY144004100MG
DSPE-PEG-Maleimide contains DSPE phospholipid and maleimide for preparing nanostructured lipid carriers. It extends blood circulation time and provides higher stability for encapsulated agents. It is for research use only and not sold to patients. Immune liposomes constructed by DSPE-PEG-Maleimide can be used for the delivery of brain drugs.
  • DSPE phospholipid and maleimide for nanostructured lipid carriers
  • Extends blood circulation time
  • Provides higher stability for encapsulated agents
  • Used for delivery of brain drugs