Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dspe-peg2000-maleimide | 00-00-0 | 99.6% | C53H99N4O14P | 10MG
SDP

Catalog No. 5000258755
Encompass_Preferred

DSPE-PEG2000-Maleimide is a phospholipid-polyethylene glycol conjugate with a terminal maleimide group that enables covalent attachment to thiol-containing ligands. It incorporates into lipid bilayers to produce targeted liposomes and lipid nanoparticles, supporting targeted delivery research and ligand-directed surface modification.

  • Terminal maleimide for specific thiol coupling.
  • Amphiphilic lipid anchor for stable liposome insertion.
  • PEG2000 spacer reduces opsonization and extends circulation time.
  • High purity suitable for research applications (>99.5%).
  • Soluble in DMF and DMSO with recommended handling procedures.
  • Storage under nitrogen at -20°C; in solvent storage at -80°C extends stability.

Catalog No. 50-002-58755 Supplier Medchemexpress LLC Supplier No. HY14073910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DSPE-PEG2000-Maleimide is a phospholipid-polyethylene glycol conjugate with a terminal maleimide group that enables covalent attachment to thiol-containing ligands. It incorporates into lipid bilayers to produce targeted liposomes and lipid nanoparticles, supporting targeted delivery research and ligand-directed surface modification.

  • Terminal maleimide for specific thiol coupling.
  • Amphiphilic lipid anchor for stable liposome insertion.
  • PEG2000 spacer reduces opsonization and extends circulation time.
  • High purity suitable for research applications (>99.5%).
  • Soluble in DMF and DMSO with recommended handling procedures.
  • Storage under nitrogen at -20°C; in solvent storage at -80°C extends stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.