Learn More
Medchemexpress LLC DSPE-PEG2000-Maleimide | 474922-22-0 | 99.58% | (C2H4O)x-C53H95NO14PNa | 100 MG
SDP
Catalog No. 5000355436
Encompass_Preferred
DSPE-PEG2000-Maleimide is a phospholipid-PEG conjugate that utilizes the amphiphilicity of DSPE to insert into the lipid bilayer of liposomes or nanoparticles. It covalently couples to the sulfhydryl (-SH) of ligands (such as antibodies, peptides, or proteins) via thiol-maleimide click chemistry, giving the particles targeting capabilities. It is for research use only and not sold to patients.
  • Utilizes DSPE amphiphilicity for lipid bilayer insertion
  • Covalently couples to sulfhydryl (-SH) of ligands
  • Enables targeting capabilities for particles
  • Used in research for breast cancer, lymphoma, and inherited retinal degeneration
  • Used to prepare targeted liposomes (TLPD)
Catalog No. 50-003-55436 Supplier Medchemexpress LLC Supplier No. HY140739100MG
DSPE-PEG2000-Maleimide is a phospholipid-PEG conjugate that utilizes the amphiphilicity of DSPE to insert into the lipid bilayer of liposomes or nanoparticles. It covalently couples to the sulfhydryl (-SH) of ligands (such as antibodies, peptides, or proteins) via thiol-maleimide click chemistry, giving the particles targeting capabilities. It is for research use only and not sold to patients.
  • Utilizes DSPE amphiphilicity for lipid bilayer insertion
  • Covalently couples to sulfhydryl (-SH) of ligands
  • Enables targeting capabilities for particles
  • Used in research for breast cancer, lymphoma, and inherited retinal degeneration
  • Used to prepare targeted liposomes (TLPD)