Learn More
Medchemexpress LLC DSPE-PEG2000-Maleimide | 99.6% | 50 MG
SDP
Catalog No. 5000356602
Encompass_Preferred
DSPE-PEG2000-Maleimide is a phospholipid-PEG conjugate. It utilizes the amphiphilicity of DSPE to insert into the lipid bilayer of liposomes or nanoparticles. This product covalently couples to the sulfhydryl (-SH) of ligands (such as antibodies, peptides, or proteins) via thiol-maleimide click chemistry, which provides targeting capabilities to the particles. It can be used in research related to breast cancer, lymphoma, and inherited retinal degeneration.
  • Used to prepare targeted liposomes (TLPD).
Catalog No. 50-003-56602 Supplier Medchemexpress LLC Supplier No. HY14073950MG
DSPE-PEG2000-Maleimide is a phospholipid-PEG conjugate. It utilizes the amphiphilicity of DSPE to insert into the lipid bilayer of liposomes or nanoparticles. This product covalently couples to the sulfhydryl (-SH) of ligands (such as antibodies, peptides, or proteins) via thiol-maleimide click chemistry, which provides targeting capabilities to the particles. It can be used in research related to breast cancer, lymphoma, and inherited retinal degeneration.
  • Used to prepare targeted liposomes (TLPD).