Learn More
Apexbio Technology LLC DSPE-PEG2000-NH₂ 474922-26-4 10mg
SDP
Catalog No. 5000097070
Encompass_Preferred
DSPE-PEG2000-NH2 (CAS 474922-26-4) is an amphiphilic polymer conjugate composed of 1 2-distearoyl-sn-glycero-3-phosphoethanolamine (DSPE) and polyethylene glycol (PEG MW 2000 Da) terminating in a primary amine The compound is designed for covalent coupling to biomolecules through its amine functional group enabling surface modification of lipid-based nanoparticles including liposomes and lipid nanoparticles DSPE-PEG2000-NH2 acts primarily by enabling conjugation of peptides proteins antibodies or nucleic acids to nanoparticle surfaces via stable amide bond formation with accessible carboxyl groups Based on these characteristics DSPE-PEG2000-NH2 holds research potential in targeted drug delivery nucleic acid delivery and nanoparticle surface modification in biomedical and pharmaceutical applications
Catalog No. 50-000-97070 Supplier Apexbio Technology LLC Supplier No. M200810
DSPE-PEG2000-NH2 (CAS 474922-26-4) is an amphiphilic polymer conjugate composed of 1 2-distearoyl-sn-glycero-3-phosphoethanolamine (DSPE) and polyethylene glycol (PEG MW 2000 Da) terminating in a primary amine The compound is designed for covalent coupling to biomolecules through its amine functional group enabling surface modification of lipid-based nanoparticles including liposomes and lipid nanoparticles DSPE-PEG2000-NH2 acts primarily by enabling conjugation of peptides proteins antibodies or nucleic acids to nanoparticle surfaces via stable amide bond formation with accessible carboxyl groups Based on these characteristics DSPE-PEG2000-NH2 holds research potential in targeted drug delivery nucleic acid delivery and nanoparticle surface modification in biomedical and pharmaceutical applications