Learn More
Medchemexpress LLC DSPE-PEG2000-PDP | 474922-24-2 | 90.0% | 25 MG
SDP
Catalog No. 5000354809
Encompass_Preferred
DSPE-PEG2000-PDP is a phospholipid PEG conjugate primarily utilized in various drug delivery applications. This compound is supplied as a white to off-white solid.
  • Phospholipid PEG conjugate
  • Suitable for drug delivery applications
  • Supplied as a white to off-white solid
  • Soluble in DMSO
Catalog No. 50-003-54809 Supplier Medchemexpress LLC Supplier No. 25MGHY144000A
DSPE-PEG2000-PDP is a phospholipid PEG conjugate primarily utilized in various drug delivery applications. This compound is supplied as a white to off-white solid.
  • Phospholipid PEG conjugate
  • Suitable for drug delivery applications
  • Supplied as a white to off-white solid
  • Soluble in DMSO