Learn More
Medchemexpress LLC DSPE-PEG2000-T7 | 3,888 Da | 10 MG
SDP
Catalog No. 5000295042
Encompass_Preferred
DSPE-PEG2000-T7 is a lipid-PEG-peptide conjugate that links a DSPE lipid to a polyethylene glycol (average Mn ~2000 Da) spacer capped with the T7 transferrin receptor-binding peptide (HAIYPRH). It is used to functionalize liposomes and nanoparticle surfaces to enable transferrin receptor-mediated targeting for drug or nucleic acid delivery; MALDI-TOF characterization reports an observed mass of ~3,888 Da.
  • Provides transferrin receptor targeting for enhanced cellular uptake.
  • Suitable for surface modification of liposomes and nanoparticles.
  • PEG spacer improves circulation and reduces nonspecific interactions.
  • Characterized by MALDI-TOF confirming expected conjugation mass.
  • Used in drug delivery and brain-targeting research applications.
Catalog No. 50-002-95042 Supplier Medchemexpress LLC Supplier No. HY17272310MG
DSPE-PEG2000-T7 is a lipid-PEG-peptide conjugate that links a DSPE lipid to a polyethylene glycol (average Mn ~2000 Da) spacer capped with the T7 transferrin receptor-binding peptide (HAIYPRH). It is used to functionalize liposomes and nanoparticle surfaces to enable transferrin receptor-mediated targeting for drug or nucleic acid delivery; MALDI-TOF characterization reports an observed mass of ~3,888 Da.
  • Provides transferrin receptor targeting for enhanced cellular uptake.
  • Suitable for surface modification of liposomes and nanoparticles.
  • PEG spacer improves circulation and reduces nonspecific interactions.
  • Characterized by MALDI-TOF confirming expected conjugation mass.
  • Used in drug delivery and brain-targeting research applications.