Learn More
Medchemexpress LLC Dspe-peg3-azide | 97.0% | 977.30 | 25 MG
SDP
Catalog No. 5000384609
Encompass_Preferred
DSPE-PEG3-azide is an azide-containing lipid that can be used to form micelles as nanoparticles for drug delivery. It functions as a click chemistry reagent, capable of undergoing copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing alkyne groups. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Can be used to form micelles for drug delivery
  • Functions as a click chemistry reagent
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reactions
  • Reacts in strain-promoted alkyne-azide cycloaddition reactions
Catalog No. 50-003-84609 Supplier Medchemexpress LLC Supplier No. 25MGHY174945
DSPE-PEG3-azide is an azide-containing lipid that can be used to form micelles as nanoparticles for drug delivery. It functions as a click chemistry reagent, capable of undergoing copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing alkyne groups. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Can be used to form micelles for drug delivery
  • Functions as a click chemistry reagent
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reactions
  • Reacts in strain-promoted alkyne-azide cycloaddition reactions