Learn More
Medchemexpress LLC DSPE-PEG3-azide | 977.30 | 5 MG
SDP
Catalog No. 5000386883
Encompass_Preferred
DSPE-PEG3-azide is an azide-containing lipid suitable for forming micelles as nanoparticles for drug delivery. It functions as a click chemistry reagent, featuring an azide group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules. It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Azide containing lipid
  • Forms micelles for drug delivery
  • Functions as a click chemistry reagent
  • Participates in CuAAc reactions
  • Undergoes SPAAC reactions
Catalog No. 50-003-86883 Supplier Medchemexpress LLC Supplier No. HY1749455MG
DSPE-PEG3-azide is an azide-containing lipid suitable for forming micelles as nanoparticles for drug delivery. It functions as a click chemistry reagent, featuring an azide group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules. It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Azide containing lipid
  • Forms micelles for drug delivery
  • Functions as a click chemistry reagent
  • Participates in CuAAc reactions
  • Undergoes SPAAC reactions