Learn More
Medchemexpress LLC DSPE-PEG4-DBCO | 2112738-14-2 | 95.0% | C71H116N3O15P | 25 MG
SDP
Catalog No. 5000352495
Encompass_Preferred
DSPE-PEG4-DBCO is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent due to its DBCO group, which facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing Azide groups. PROTACs themselves are composed of two distinct ligands connected by a linker: one ligand binds to an E3 ubiquitin ligase, and the other targets a specific protein. These PROTACs leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • Peg-based PROTAC linker
  • Click chemistry reagent
  • Contains a DBCO group for strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules
  • Used in the synthesis of PROTACs
  • PROTACs utilize the ubiquitin-proteasome system for selective protein degradation
Catalog No. 50-003-52495 Supplier Medchemexpress LLC Supplier No. 25MGHY140955
DSPE-PEG4-DBCO is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent due to its DBCO group, which facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing Azide groups. PROTACs themselves are composed of two distinct ligands connected by a linker: one ligand binds to an E3 ubiquitin ligase, and the other targets a specific protein. These PROTACs leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • Peg-based PROTAC linker
  • Click chemistry reagent
  • Contains a DBCO group for strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules
  • Used in the synthesis of PROTACs
  • PROTACs utilize the ubiquitin-proteasome system for selective protein degradation