Learn More
Medchemexpress LLC Dspe-peg4-dbco | 2112738-14-2 | MFCD30828684 | 95.0% | C71H116N3O15P | 10MG
SDP
Catalog No. 5000260470
Encompass_Preferred
DSPE-PEG4-DBCO is a lipid-polyethylene glycol conjugate bearing a dibenzocyclooctyne (DBCO) reactive group used for copper-free click chemistry and bioconjugation. It enables strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-functionalized partners, facilitating liposome functionalization, surface modification, and linker assembly in aqueous and physiological conditions.
  • Provides a DBCO reactive handle for copper-free SPAAC.
  • Contains a PEG4 spacer for improved solubility and reduced steric hindrance.
  • Includes a lipid anchor for incorporation into lipid bilayers and liposomes.
  • Suitable for bioconjugation under physiological conditions.
  • High purity for consistent reaction performance.
Catalog No. 50-002-60470 Supplier Medchemexpress LLC Supplier No. HY14095510MG
DSPE-PEG4-DBCO is a lipid-polyethylene glycol conjugate bearing a dibenzocyclooctyne (DBCO) reactive group used for copper-free click chemistry and bioconjugation. It enables strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-functionalized partners, facilitating liposome functionalization, surface modification, and linker assembly in aqueous and physiological conditions.
  • Provides a DBCO reactive handle for copper-free SPAAC.
  • Contains a PEG4 spacer for improved solubility and reduced steric hindrance.
  • Includes a lipid anchor for incorporation into lipid bilayers and liposomes.
  • Suitable for bioconjugation under physiological conditions.
  • High purity for consistent reaction performance.