Learn More
Medchemexpress LLC DSPE-PEG4-DBCO | 2112738-14-2 | MFCD30828684 | 95.0% | C71H116N3O15P | 50 MG
SDP
Catalog No. 5000355076
Encompass_Preferred
DSPE-PEG4-DBCO is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It functions as a click chemistry reagent, incorporating a DBCO group that facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing Azide groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains a DBCO group
  • Facilitates strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Reacts with molecules containing azide groups
Catalog No. 50-003-55076 Supplier Medchemexpress LLC Supplier No. HY14095550MG
DSPE-PEG4-DBCO is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It functions as a click chemistry reagent, incorporating a DBCO group that facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing Azide groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains a DBCO group
  • Facilitates strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Reacts with molecules containing azide groups