Learn More
Medchemexpress LLC Dspe-peg5000-mal ammonium | 474922-22-0 | 98.4% | (C2H4O)nC51H92N1O13P.H3N | 25 MG
SDP
Catalog No. 5000354810
Encompass_Preferred
DSPE-PEG-Maleimide has DSPE phospholipid and maleimide to prepare nanostructured lipid carrier. DSPE-PEG-Maleimide extends blood circulation time and higher stability for encapsulated agents. DSPE-PEG5000-Mal ammonium contains PEG5000.
  • Contains DSPE phospholipid and maleimide for nanostructured lipid carrier preparation
  • Extends blood circulation time
  • Provides higher stability for encapsulated agents
  • Contains PEG5000
Catalog No. 50-003-54810 Supplier Medchemexpress LLC Supplier No. 25MGHY144004A
DSPE-PEG-Maleimide has DSPE phospholipid and maleimide to prepare nanostructured lipid carrier. DSPE-PEG-Maleimide extends blood circulation time and higher stability for encapsulated agents. DSPE-PEG5000-Mal ammonium contains PEG5000.
  • Contains DSPE phospholipid and maleimide for nanostructured lipid carrier preparation
  • Extends blood circulation time
  • Provides higher stability for encapsulated agents
  • Contains PEG5000