Learn More
Medchemexpress LLC Dspe-peg5000-mal ammonium | 474922-22-0 | 98.4% | (C2H4O)nC51H92N1O13P.H3N | 50 MG
SDP
Catalog No. 5000356164
Encompass_Preferred
DSPE-PEG5000-Mal ammonium contains DSPE phospholipid and maleimide, making it suitable for preparing nanostructured lipid carriers. It is designed to enhance blood circulation time and provide higher stability for encapsulated agents. This product specifically contains PEG5000 and can be utilized in constructing immune liposomes for the delivery of brain drugs.
  • Used to prepare nanostructured lipid carriers
  • Enhances blood circulation time
  • Provides higher stability for encapsulated agents
  • Suitable for brain drug delivery
Catalog No. 50-003-56164 Supplier Medchemexpress LLC Supplier No. HY144004A50MG
DSPE-PEG5000-Mal ammonium contains DSPE phospholipid and maleimide, making it suitable for preparing nanostructured lipid carriers. It is designed to enhance blood circulation time and provide higher stability for encapsulated agents. This product specifically contains PEG5000 and can be utilized in constructing immune liposomes for the delivery of brain drugs.
  • Used to prepare nanostructured lipid carriers
  • Enhances blood circulation time
  • Provides higher stability for encapsulated agents
  • Suitable for brain drug delivery