Learn More
Medchemexpress LLC DSPE-PEG6-Mal | 95.0% | 1163.50 | 100 MG
SDP
Catalog No. 5000350292
Encompass_Preferred
DSPE-PEG6-Mal is a versatile reagent used in various biochemical and biomedical applications. It is commonly employed in drug delivery systems, particularly for modifying liposomes and nanoparticles to improve their circulation half-life and targeting capabilities. This compound facilitates bioconjugation reactions due to its maleimide group, which readily reacts with thiol-containing molecules.
  • Facilitates bioconjugation reactions
  • Used for surface modification of nanoparticles
  • Enhances drug delivery systems
  • Enables protein and peptide modification
Catalog No. 50-003-50292 Supplier Medchemexpress LLC Supplier No. HY138395100MG
DSPE-PEG6-Mal is a versatile reagent used in various biochemical and biomedical applications. It is commonly employed in drug delivery systems, particularly for modifying liposomes and nanoparticles to improve their circulation half-life and targeting capabilities. This compound facilitates bioconjugation reactions due to its maleimide group, which readily reacts with thiol-containing molecules.
  • Facilitates bioconjugation reactions
  • Used for surface modification of nanoparticles
  • Enhances drug delivery systems
  • Enables protein and peptide modification