Learn More
Medchemexpress LLC DSPE-PEG6-Mal | 95.0% | 1163.50 | 50 MG
SDP
Catalog No. 5000351123
Encompass_Preferred
DSPE-PEG6-Mal is a PEG-based PROTAC linker primarily used in the synthesis of PROTACs. PROTACs are designed with two ligands connected by a linker: one targeting an E3 ubiquitin ligase and the other for a specific target protein. This mechanism utilizes the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is strictly for research use and is not intended for medical, clinical diagnostic, or food applications in humans or animals.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates targeted protein degradation via the ubiquitin-proteasome system
  • White to off-white solid appearance
  • Purity of 95.0%
Catalog No. 50-003-51123 Supplier Medchemexpress LLC Supplier No. HY13839550MG
DSPE-PEG6-Mal is a PEG-based PROTAC linker primarily used in the synthesis of PROTACs. PROTACs are designed with two ligands connected by a linker: one targeting an E3 ubiquitin ligase and the other for a specific target protein. This mechanism utilizes the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is strictly for research use and is not intended for medical, clinical diagnostic, or food applications in humans or animals.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates targeted protein degradation via the ubiquitin-proteasome system
  • White to off-white solid appearance
  • Purity of 95.0%