Learn More
Medchemexpress LLC DSPE-pSar50 (DSPE-Polysarcosine50) | 3091879-29-4 | 99.91% | 4319.06 | 5 MG
SDP
Catalog No. 5000390904
Encompass_Preferred
DSPE-pSar50 (DSPE-Polysarcosine50) is a pSar-lipid derivative that serves as a hydrophilic alternative to PEG. It can be utilized in drug delivery research. The product is for research use only and not sold to patients.
  • pSar-lipid derivative
  • Hydrophilic alternative to PEG
  • Utilized in drug delivery research
  • For research use only
Catalog No. 50-003-90904 Supplier Medchemexpress LLC Supplier No. HY1670645MG
DSPE-pSar50 (DSPE-Polysarcosine50) is a pSar-lipid derivative that serves as a hydrophilic alternative to PEG. It can be utilized in drug delivery research. The product is for research use only and not sold to patients.
  • pSar-lipid derivative
  • Hydrophilic alternative to PEG
  • Utilized in drug delivery research
  • For research use only