Learn More
Medchemexpress LLC Dspe-pSar50 (dspe-polysarcosine50) | 99.91% | 1 MG
SDP
Catalog No. 5000390939
Encompass_Preferred
DSPE-pSar50 (DSPE-Polysarcosine50) is a pSar-lipid derivative. It is a hydrophilic alternative to PEG and can be used in drug delivery research.
  • PSar-lipid derivative
  • Hydrophilic alternative to PEG
  • Can be used in drug delivery research
Catalog No. 50-003-90939 Supplier Medchemexpress LLC Supplier No. HY1670641MG
DSPE-pSar50 (DSPE-Polysarcosine50) is a pSar-lipid derivative. It is a hydrophilic alternative to PEG and can be used in drug delivery research.
  • PSar-lipid derivative
  • Hydrophilic alternative to PEG
  • Can be used in drug delivery research