Learn More
Medchemexpress LLC DSPE-succinic acid | 248253-94-3 | 98.0% | 848.1 g/mol | C45H86NO11P | 100 MG
SDP
Catalog No. 5000281409
Encompass_Preferred
DSPE-succinic acid is a phospholipid bearing a terminal carboxylic acid moiety that enables covalent conjugation to amine-containing molecules. It is used to functionalize lipids for the preparation of nanoparticles and liposomes, facilitating attachment of targeting ligands or cargo for therapeutic delivery.
  • Terminal carboxylic acid for conjugation to amines.
  • Forms stable amide bonds with primary amines.
  • Suitable for liposome and nanoparticle formulation.
  • High purity suitable for research use (98.0%).
  • Soluble in DMF for formulation and conjugation reactions.
Catalog No. 50-002-81409 Supplier Medchemexpress LLC Supplier No. HYW441002100MG
DSPE-succinic acid is a phospholipid bearing a terminal carboxylic acid moiety that enables covalent conjugation to amine-containing molecules. It is used to functionalize lipids for the preparation of nanoparticles and liposomes, facilitating attachment of targeting ligands or cargo for therapeutic delivery.
  • Terminal carboxylic acid for conjugation to amines.
  • Forms stable amide bonds with primary amines.
  • Suitable for liposome and nanoparticle formulation.
  • High purity suitable for research use (98.0%).
  • Soluble in DMF for formulation and conjugation reactions.