Learn More
Medchemexpress LLC DSPE-succinic acid | 248253-94-3 | 98.0% | 848.14 g/mol | C45H86NO11P | 25 MG
SDP
Catalog No. 5000281140
Encompass_Preferred
DSPE-succinic acid is a phospholipid bearing a terminal succinic (carboxylic acid) group that enables covalent conjugation to amine-containing ligands. It is commonly used for preparing liposomes and nanoparticles and for surface functionalization in drug delivery and bioconjugation workflows.
  • Terminal carboxylic acid enables stable amide bond formation with amines.
  • Suitable for liposome and nanoparticle formulation.
  • Facilitates surface functionalization for targeting or PEGylation.
  • High purity (98.0%) appropriate for research applications.
  • Available in small, research-scale quantities for formulation work.
Catalog No. 50-002-81140 Supplier Medchemexpress LLC Supplier No. HYW44100225MG
DSPE-succinic acid is a phospholipid bearing a terminal succinic (carboxylic acid) group that enables covalent conjugation to amine-containing ligands. It is commonly used for preparing liposomes and nanoparticles and for surface functionalization in drug delivery and bioconjugation workflows.
  • Terminal carboxylic acid enables stable amide bond formation with amines.
  • Suitable for liposome and nanoparticle formulation.
  • Facilitates surface functionalization for targeting or PEGylation.
  • High purity (98.0%) appropriate for research applications.
  • Available in small, research-scale quantities for formulation work.