Learn More
Medchemexpress LLC DSPE-succinic acid | 248253-94-3 | 98.0% | 848.14 g·mol⁻¹ | C45H86NO11P | 5 MG
SDP
Catalog No. 5000280972
Encompass_Preferred
DSPE-succinic acid is a phospholipid derivative bearing a terminal carboxylic acid that enables covalent coupling to amine-containing molecules. It is used as a functional lipid in liposomes and nanoparticle formulations to provide reactive surface functionality for bioconjugation and targeted delivery applications.
  • Terminal carboxylic acid enables amide bond formation with amines.
  • Suitable for liposome and nanoparticle formulation and surface functionalization.
  • High purity (98.0%) for consistent performance in conjugation reactions.
  • Available in small sample sizes for research use, including 5 MG.
  • Stable as powder at -20 °C for long-term storage.
Catalog No. 50-002-80972 Supplier Medchemexpress LLC Supplier No. HYW4410025MG
DSPE-succinic acid is a phospholipid derivative bearing a terminal carboxylic acid that enables covalent coupling to amine-containing molecules. It is used as a functional lipid in liposomes and nanoparticle formulations to provide reactive surface functionality for bioconjugation and targeted delivery applications.
  • Terminal carboxylic acid enables amide bond formation with amines.
  • Suitable for liposome and nanoparticle formulation and surface functionalization.
  • High purity (98.0%) for consistent performance in conjugation reactions.
  • Available in small sample sizes for research use, including 5 MG.
  • Stable as powder at -20 °C for long-term storage.