Learn More
Medchemexpress LLC DSPE-succinic acid | 248253-94-3 | 98.0% | 848.14 g/mol | C45H86NO11P | 50 MG
SDP
Catalog No. 5000281767
Encompass_Preferred
DSPE-succinic acid is a phospholipid bearing a terminal carboxylic acid group. The acid functionality enables covalent coupling to amines via amide bond formation, making the compound useful for surface functionalization and bioconjugation in liposome and nanoparticle formulations for drug delivery research.
  • Carboxylic acid functionality reactive toward amines for stable amide conjugation.
  • Suitable for liposome and nanoparticle formulation and surface functionalization.
  • Facilitates bioconjugation of ligands, peptides, or polymers.
  • High purity for research applications (reported 98.0%).
  • Amphiphilic structure supports incorporation into lipid bilayers.
Catalog No. 50-002-81767 Supplier Medchemexpress LLC Supplier No. HYW44100250MG
DSPE-succinic acid is a phospholipid bearing a terminal carboxylic acid group. The acid functionality enables covalent coupling to amines via amide bond formation, making the compound useful for surface functionalization and bioconjugation in liposome and nanoparticle formulations for drug delivery research.
  • Carboxylic acid functionality reactive toward amines for stable amide conjugation.
  • Suitable for liposome and nanoparticle formulation and surface functionalization.
  • Facilitates bioconjugation of ligands, peptides, or polymers.
  • High purity for research applications (reported 98.0%).
  • Amphiphilic structure supports incorporation into lipid bilayers.