Learn More
Medchemexpress LLC DSPE-thiol | 144735-82-0 | 90.0% | 836.19 g/mol | C44H86NO9PS | 10 MG
SDP
Catalog No. 5000280973
Encompass_Preferred
DSPE-thiol is a distearoylphosphatidylethanolamine lipid functionalized with a terminal thiol (-SH) group. It introduces reactive thiol chemistry to lipid assemblies, enabling covalent linkage to maleimide-containing molecules and the creation of phospholipid dimers for nanocarrier and membrane research.
  • Provides reactive thiol functionality for selective conjugation with maleimide reagents.
  • Facilitates liposome surface functionalization and targeted delivery studies.
  • Enables synthesis of phospholipid dimers and cross-linked lipid structures.
  • Integrates into lipid bilayers to impart stable, hydrophobic anchoring.
  • Supplied in small research-scale quantities for laboratory use.
Catalog No. 50-002-80973 Supplier Medchemexpress LLC Supplier No. HYW44100410MG
DSPE-thiol is a distearoylphosphatidylethanolamine lipid functionalized with a terminal thiol (-SH) group. It introduces reactive thiol chemistry to lipid assemblies, enabling covalent linkage to maleimide-containing molecules and the creation of phospholipid dimers for nanocarrier and membrane research.
  • Provides reactive thiol functionality for selective conjugation with maleimide reagents.
  • Facilitates liposome surface functionalization and targeted delivery studies.
  • Enables synthesis of phospholipid dimers and cross-linked lipid structures.
  • Integrates into lipid bilayers to impart stable, hydrophobic anchoring.
  • Supplied in small research-scale quantities for laboratory use.