Learn More
Kyfora Bio DSPE 250 MG
Catalog No. 502559770
Encompass_Preferred
DSPE (1,2-Distearoyl-rac-glycero-3-Phosphatidylethanolamine) is a saturated zwitterionic phospholipid and a derivative of phosphatidylethanolamine. It can be used in liposomal drug and gene delivery formulations. DSPE can be conjugated with PEG or PCB to develop stealth liposomes that have extended circulation time in the bloodstream.Key application(s): LNP, stealth liposomes
Catalog No. 50-255-9770 Supplier Kyfora Bio Supplier No. 26452250
DSPE (1,2-Distearoyl-rac-glycero-3-Phosphatidylethanolamine) is a saturated zwitterionic phospholipid and a derivative of phosphatidylethanolamine. It can be used in liposomal drug and gene delivery formulations. DSPE can be conjugated with PEG or PCB to develop stealth liposomes that have extended circulation time in the bloodstream.Key application(s): LNP, stealth liposomes